Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042399.1 | NP_001035858.1 | 7 | 1402 | ||
NM_016122.2 | NP_057206.2 | 8 | 1455 | |||
XM_006719437.2 | XP_006719500.1 | 7 | 1368 | |||
XM_011538424.1 | XP_011536726.1 | 9 | 1501 | |||
XM_011538427.2 | XP_011536729.1 | 8 | 1227 | |||
XM_011538429.2 | XP_011536731.1 | 4 | 698 | |||
XM_017019385.1 | XP_016874874.1 | 8 | 1449 | |||
XM_017019386.1 | XP_016874875.1 | 8 | 1449 | |||
XM_017019387.1 | XP_016874876.1 | 7 | 1206 | |||
XM_017019388.1 | XP_016874877.1 | 7 | 3071 | |||
XM_017019389.1 | XP_016874878.1 | 9 | 1281 | |||
GenBank mRNA | AF155115.1 | - | 8 | 1171 | ||
AK056316.1 | - | 7 | 1298 | |||
BC107746.1 | - | 5 | 716 | |||
BC125086.1 | - | 7 | 1092 | |||
BC125087.1 | - | 8 | 1005 | 40117807 |