Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INSIG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321329.1 | NP_001308258.1 | 4 | 638 | ||
NM_001321330.1 | NP_001308259.1 | 3 | 263 | |||
NM_001321331.1 | NP_001308260.1 | 4 | 271 | |||
NM_001321332.1 | NP_001308261.1 | 4 | 264 | |||
NM_001321333.1 | NP_001308262.1 | 4 | 271 | |||
NM_016133.3 | NP_057217.2 | 4 | 645 | |||
GenBank mRNA | AF125392.1 | - | 3 | 559 | ||
AF527632.1 | - | 4 | 594 | |||
AK021692.1 | - | 4 | 176 | |||
AK291433.1 | - | 4 | 593 | |||
AK298599.1 | - | 3 | 227 | |||
BC022475.1 | - | 4 | 589 | 4794938 | ||
BX647805.1 | - | 4 | 186 | |||
DC364733.1 | - | 3 | 228 |