Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INSIG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321329.1 | NP_001308258.1 | 5 | 872 | ||
NM_001321330.1 | NP_001308259.1 | 4 | 497 | |||
NM_001321331.1 | NP_001308260.1 | 5 | 505 | |||
NM_001321332.1 | NP_001308261.1 | 5 | 498 | |||
NM_016133.3 | NP_057217.2 | 5 | 879 | |||
GenBank mRNA | AF125392.1 | - | 4 | 793 | ||
AF527632.1 | - | 5 | 828 | |||
AK291433.1 | - | 5 | 827 | |||
AK298599.1 | - | 4 | 461 | |||
AL080184.1 | - | 1 | 14 | |||
BC022475.1 | - | 5 | 823 | 4794938 | ||
BE888848.1 | - | 5 | 504 | |||
BX647805.1 | - | 5 | 420 | |||
DC364733.1 | - | 4 | 462 |