Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD17B12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016142.2 | NP_057226.1 | 3 | 463 | ||
XM_011520156.1 | XP_011518458.1 | 3 | 457 | |||
XM_017017881.1 | XP_016873370.1 | 3 | 143 | |||
GenBank mRNA | AF078850.1 | - | 3 | 301 | ||
AK027882.1 | - | 357 | ||||
AK074952.1 | - | 3 | 315 | |||
AK075011.1 | - | 3 | 348 | |||
AK075216.1 | - | 3 | 324 | |||
AK222881.1 | - | 3 | 285 | |||
AK292625.1 | - | 3 | 348 | |||
AK301663.1 | - | 4 | 254 | |||
BC012043.1 | - | 3 | 286 | 3622879 | ||
BC012536.1 | - | 3 | 279 |