Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD17B12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016142.2 | NP_057226.1 | 8 | 842 | ||
XM_011520156.1 | XP_011518458.1 | 8 | 836 | |||
XM_017017881.1 | XP_016873370.1 | 8 | 522 | |||
GenBank mRNA | AF078850.1 | - | 8 | 680 | ||
AK027882.1 | - | 735 | ||||
AK074952.1 | - | 8 | 694 | |||
AK075011.1 | - | 8 | 727 | |||
AK075216.1 | - | 8 | 703 | |||
AK092938.1 | - | 1019 | ||||
AK222881.1 | - | 8 | 664 | |||
AK292625.1 | - | 8 | 727 | |||
AK301663.1 | - | 9 | 633 | |||
BC012043.1 | - | 8 | 665 | 3622879 |