Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C5orf45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017987.2 | NP_001017987.1 | 2 | 155 | ||
NM_016175.3 | NP_057259.2 | 2 | 155 | |||
GenBank mRNA | AK293901.1 | - | 2 | 121 | ||
AK295104.1 | - | 2 | 148 | |||
AK298954.1 | - | 2 | 112 | |||
BC056889.1 | - | 2 | 108 | |||
BC069051.1 | - | 2 | 123 | 3945859 | ||
BF965313.1 | - | 2 | 156 | |||
BQ642639.1 | - | 2 | 111 | |||
BX537968.1 | - | 2 | 176 | |||
BX647375.1 | - | 2 | 123 | |||
CN363239.1 | - | 2 | 79 | |||
CR936785.1 | - | 2 | 176 |