Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C5orf45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017987.2 | NP_001017987.1 | 3 | 315 | ||
NM_016175.3 | NP_057259.2 | 5 | 480 | |||
GenBank mRNA | AF153685.1 | - | 1 | 101 | ||
AK293901.1 | - | 5 | 446 | |||
AK295104.1 | - | 3 | 308 | |||
AK298954.1 | - | 3 | 272 | |||
BC002739.2 | - | 1 | 27 | |||
BC056889.1 | - | 4 | 344 | |||
BC069051.1 | - | 5 | 448 | 3945859 | ||
BF965313.1 | - | 4 | 404 | |||
BQ642639.1 | - | 3 | 271 | |||
BX537968.1 | - | 6 | 581 | |||
BX647375.1 | - | 5 | 448 | |||
CN363239.1 | - | 6 | 484 |