Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC45A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012509.3 | NP_001012527.1 | 1, 2 | 485 | ||
NM_001297417.2 | NP_001284346.2 | 1, 2 | 485 | |||
NM_016180.4 | NP_057264.3 | 1, 2 | 485 | |||
GenBank mRNA | AF172849.1 | - | 1, 2 | 418 | ||
BC003597.1 | - | 1, 2 | 454 | |||
BC064405.1 | - | 1, 2 | 451 | 5563039 | ||
BQ677785.1 | - | 1, 2 | 485 |