Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002032.2 | NP_001002032.1 | 4 | 1374 | ||
NM_001002033.2 | NP_001002033.1 | 6 | 1488 | |||
NM_001288609.1 | NP_001275538.1 | 5 | 2077 | |||
NM_001288610.1 | NP_001275539.1 | 5 | 1440 | |||
NM_001288611.1 | NP_001275540.1 | 5 | 1397 | |||
NM_016185.3 | NP_057269.1 | 5 | 1393 | |||
NR_109933.1 | 5 | 1429 | ||||
GenBank mRNA | AF060925.1 | - | 1 | 659 | ||
AF086910.2 | - | 5 | 1393 | |||
AF266846.1 | - | 5 | 1222 | |||
AK223321.1 | - | 5 | 1222 | |||
AK308253.1 | - | 5 | 1439 | |||
AY322169.1 | - | 4 | 1366 | |||
AY322170.1 | - | 6 | 1480 | |||
BC039343.1 | - | 5 | 2079 | 5269120 |