Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016208.3 | NP_057292.1 | 3 | 161 | ||
NM_183057.2 | NP_898880.1 | 3 | 161 | |||
XM_005272323.3 | XP_005272380.1 | 3 | 153 | |||
XM_005272324.3 | XP_005272381.1 | 3 | 153 | |||
XM_005272325.3 | XP_005272382.1 | 3 | 146 | |||
XM_011517099.1 | XP_011515401.1 | 3 | 111 | |||
XM_011517100.1 | XP_011515402.1 | 3 | 111 | |||
XM_017013529.1 | XP_016869018.1 | 3 | 146 | |||
GenBank mRNA | AF182844.1 | - | 2 | 113 | ||
AF316887.1 | - | 2 | 95 | |||
AK026865.1 | - | 3 | 111 | |||
AK090942.1 | - | 3 | 111 | |||
AK091979.1 | - | 3 | 111 | |||
AK310062.1 | - | 3 | 153 | |||
BC006485.1 | - | 2 | 97 | 3535787 | ||
BC019321.1 | - | 2 | 97 | 2822758 | ||
BC050713.2 | - | 3 | 109 | 5741129 | ||
BP380505.1 | - | 3 | 164 | |||
BU658837.1 | - | 3 | 454 | |||
KU178635.1 | - | 2 | 52 | |||
KU178636.1 | - | 2 | 52 | |||
R72904.1 | - | 2 | 43 |