Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDC25C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287582.1 | NP_001274511.1 | 2 | 465 | ||
NM_001287583.1 | NP_001274512.1 | 2 | 582 | |||
NM_001318098.1 | NP_001305027.1 | 2 | 586 | |||
NM_001790.4 | NP_001781.2 | 2 | 469 | |||
NM_022809.3 | NP_073720.1 | 2 | 469 | |||
XM_005272145.3 | XP_005272202.1 | 2 | 582 | |||
XM_011543759.2 | XP_011542061.1 | 3 | 253 | |||
XM_011543760.2 | XP_011542062.1 | 2 | 582 | |||
XM_011543761.2 | XP_011542063.1 | 2 | 582 | |||
XM_011543762.1 | XP_011542064.1 | 2 | 382 | |||
XM_011543764.1 | XP_011542066.1 | 1 | 55 | |||
XM_017010101.1 | XP_016865590.1 | 2 | 460 | |||
GenBank mRNA | AF277723.1 | - | 1 | 55 | ||
AF277724.1 | - | 1 | 55 | |||
AF277725.1 | - | 1 | 55 | |||
AF277726.1 | - | 1 | 55 | |||
AJ304504.1 | - | 1 | 148 | |||
AK097710.1 | - | 2 | 582 | |||
AK301828.1 | - | 3 | 532 | |||
AU125025.1 | - | 2 | 390 | |||
BC019089.2 | - | 2 | 359 | 4858949 | ||
DB040755.1 | - | 2 | 390 | |||
M34065.1 | - | 2 | 358 |