Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUBE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016262.4 | NP_057346.1 | 7 | 675 | ||
XM_011535875.2 | XP_011534177.1 | 6 | 598 | |||
XM_017010918.1 | XP_016866407.1 | 8 | 689 | |||
GenBank mRNA | AF201334.1 | - | 7 | 561 | ||
AK026242.1 | - | 7 | 640 | |||
AK126191.1 | - | 1 | 2297 | |||
BC025405.2 | - | 7 | 580 | 4824105 | ||
BC031101.1 | - | 7 | 628 | 5298159 |