Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRBN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173482.1 | NP_001166953.1 | 6, 7 | 783 | ||
NM_016302.3 | NP_057386.2 | 6, 7 | 786 | |||
XM_005265202.3 | XP_005265259.1 | 5, 6 | 667 | |||
XM_011533791.2 | XP_011532093.1 | 6, 7 | 773 | |||
XM_011533793.2 | XP_011532095.1 | 7, 8 | 914 | |||
XM_011533794.2 | XP_011532096.1 | 6, 7 | 785 | |||
GenBank mRNA | AF117230.1 | - | 6, 7 | 750 | ||
AK027507.1 | - | 6, 7 | 764 | |||
AK058191.1 | - | 6, 7 | 760 | |||
AK301423.1 | - | 5, 6 | 667 | |||
AK312577.1 | - | 6, 7 | 764 | |||
BC017419.1 | - | 6, 7 | 774 | 4672890 | ||
BC067811.1 | - | 7, 8 | 767 | 5264951 |