Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRBN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173482.1 | NP_001166953.1 | 4 | 479 | ||
NM_016302.3 | NP_057386.2 | 4 | 482 | |||
XM_005265202.3 | XP_005265259.1 | 3 | 363 | |||
XM_011533791.2 | XP_011532093.1 | 4 | 469 | |||
XM_011533793.2 | XP_011532095.1 | 4 | 469 | |||
XM_011533794.2 | XP_011532096.1 | 3 | 340 | |||
GenBank mRNA | AF117230.1 | - | 4 | 446 | ||
AK027507.1 | - | 4 | 460 | |||
AK058191.1 | - | 4 | 456 | |||
AK301423.1 | - | 3 | 363 | |||
AK312577.1 | - | 4 | 460 | |||
BC017419.1 | - | 4 | 470 | 4672890 | ||
BC067811.1 | - | 4 | 463 | 5264951 | ||
CR627060.1 | - | 3 | 349 | |||
DB456585.1 | - | 4 | 483 |