Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SS18L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016305.3 | NP_057389.1 | 4 | 373 | ||
XM_017006548.1 | XP_016862037.1 | 3 | 1385 | |||
XM_017006549.1 | XP_016862038.1 | 4 | 344 | |||
GenBank mRNA | AF201950.1 | - | 3 | 263 | ||
AK312225.1 | - | 3 | 168 | |||
BC017804.1 | - | 3 | 191 | 4658643 | ||
BC020444.1 | - | 3 | 163 | |||
BX114027.1 | - | 4 | 367 |