Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HERC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016323.3 | NP_057407.2 | 14 | 1904 | ||
GenBank mRNA | AB027289.1 | - | 14 | 1864 | ||
AK093155.1 | - | 4 | 1239 | |||
AK301756.1 | - | 4 | 472 | |||
AK302142.1 | - | 8 | 776 | |||
AY337518.1 | - | 14 | 1904 | |||
BC140716.1 | - | 14 | 1797 | 9021584 |