Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IPO11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134779.1 | NP_001128251.1 | 22 | 2289 | ||
NM_016338.4 | NP_057422.3 | 22 | 2229 | |||
GenBank mRNA | AF111109.1 | - | 21 | 2039 | ||
AK001696.1 | - | 22 | 2211 | |||
AK023177.1 | - | 10 | 802 | |||
AK302781.1 | - | 22 | 2289 | |||
BC031694.1 | - | 14 | 1290 | |||
BC033776.1 | - | 22 | 2191 | 4525238 |