Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAPGEFL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303533.1 | NP_001290462.1 | 11 | 1227 | ||
NM_001303534.1 | NP_001290463.1 | 11 | 1322 | |||
NM_016339.4 | NP_057423.1 | 11 | 1507 | |||
XM_006721932.3 | XP_006721995.1 | 11 | 2001 | |||
XM_006721935.3 | XP_006721998.1 | 12 | 1278 | |||
XM_006721937.3 | XP_006722000.1 | 11 | 3103 | |||
XM_006721938.3 | XP_006722001.1 | 8 | 894 | |||
GenBank mRNA | AF117946.1 | - | 11 | 1142 | ||
AK294642.1 | - | 7 | 1129 | |||
AK304160.1 | - | 11 | 1322 | |||
AK313492.1 | - | 11 | 1182 | |||
BC115374.1 | - | 9 | 1036 | 40028278 | ||
BC115375.1 | - | 9 | 1036 | 40028280 |