Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016370.2 | NP_057454.1 | 3 | 815 | ||
GenBank mRNA | AB036693.1 | - | 3 | 761 | ||
AK313425.1 | - | 3 | 710 | |||
BC093756.1 | - | 1 | 499 | 7939601 | ||
BC093758.1 | - | 1 | 499 | 7939603 |