Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LUZP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318840.1 | NP_001305769.1 | 3 | 404 | ||
NM_016383.4 | NP_057467.1 | 4 | 523 | |||
GenBank mRNA | AF124430.1 | - | 4 | 493 | ||
AK093369.1 | - | 3 | 395 | |||
AK314765.1 | - | 4 | 512 | |||
BC128134.1 | - | 4 | 485 | 40112435 | ||
BC128135.1 | - | 4 | 484 | |||
DB041564.1 | - | 4 | 515 |