Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF219 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101672.1 | NP_001095142.1 | 3 | 501 | ||
NM_001102454.1 | NP_001095924.1 | 3 | 491 | |||
NM_016423.2 | NP_057507.2 | 3 | 635 | |||
XM_005267739.1 | XP_005267796.1 | 3 | 446 | |||
XM_006720163.2 | XP_006720226.1 | 3 | 382 | |||
XM_006720164.3 | XP_006720227.1 | 3 | 384 | |||
XM_011536825.1 | XP_011535127.1 | 3 | 456 | |||
XM_017021354.1 | XP_016876843.1 | 2 | 451 | |||
XM_017021355.1 | XP_016876844.1 | 3 | 347 | |||
GenBank mRNA | AB015427.1 | - | 3 | 625 | ||
AK225811.1 | - | 3 | 398 | |||
AK290700.1 | - | 3 | 501 | |||
BC000694.2 | - | 2 | 252 | 3347777 | ||
BC036105.1 | - | 3 | 327 | 5274736 | ||
BT006956.1 | - | 1 | 214 | |||
CB152705.1 | - | 4 | 465 | |||
DA013397.1 | - | 3 | 491 |