Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCEB3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016427.2 | NP_057511.2 | 1 | 2108 | ||
GenBank mRNA | AB030834.1 | - | 1 | 1869 | ||
AK314984.1 | - | 1 | 2088 | |||
BC036022.1 | - | 1 | 2112 | 5270406 | ||
BC103911.2 | - | 1 | 1756 | 40006025 |