Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIGP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320480.1 | NP_001307409.1 | 4 | 588 | ||
NM_016430.3 | NP_057514.2 | 5 | 602 | |||
NM_153681.2 | NP_710148.1 | 3 | 543 | |||
NM_153682.2 | NP_710149.1 | 4 | 436 | |||
XM_005260990.4 | XP_005261047.1 | 4 | 301 | |||
XM_011529596.2 | XP_011527898.1 | 4 | 335 | |||
XM_017028364.1 | XP_016883853.1 | 4 | 353 | |||
XM_017028365.1 | XP_016883854.1 | 5 | 544 | |||
GenBank mRNA | AB035742.1 | - | 4 | 368 | ||
AB035743.1 | - | 4 | 298 | |||
AB035744.1 | - | 4 | 330 | |||
AB035745.1 | - | 3 | 350 | |||
AB037162.1 | - | 4 | 372 | |||
AB037163.1 | - | 3 | 341 | |||
AB037164.1 | - | 4 | 453 | |||
AB039659.1 | - | 3 | 255 | |||
AB451328.1 | - | 2 | 177 | |||
AB451472.1 | - | 2 | 177 | |||
AF216305.1 | - | 5 | 518 | |||
AF237812.1 | - | 3 | 392 | |||
AK026311.1 | - | 3 | 328 | |||
AK304656.1 | - | 3 | 462 | |||
AK314457.1 | - | 3 | 336 | |||
AK316609.1 | - | 3 | 273 | |||
BC005180.1 | - | 4 | 317 | 3929930 | ||
BC011007.1 | - | 4 | 321 | 4281552 | ||
BE275469.1 | - | 4 | 435 | |||
BT007053.1 | - | 3 | 255 |