Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHF20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016436.4 | NP_057520.2 | 5, 6 | 539 | ||
XM_005260421.2 | XP_005260478.1 | 2, 3 | 149 | |||
XM_011528844.1 | XP_011527146.1 | 2, 3 | 211 | |||
XM_017027864.1 | XP_016883353.1 | 6, 7 | 621 | |||
XM_017027865.1 | XP_016883354.1 | 6, 7 | 822 | |||
XM_017027866.1 | XP_016883355.1 | 5, 6 | 742 | |||
XM_017027867.1 | XP_016883356.1 | 5, 6 | 577 | |||
XM_017027871.1 | XP_016883360.1 | 6, 7 | 622 | |||
GenBank mRNA | AF220416.1 | - | 5, 6 | 503 | ||
AF258787.1 | - | 5, 6 | 525 | |||
AF348207.1 | - | 4, 5 | 464 | |||
AK054597.1 | - | 6, 7 | 796 | |||
AK314503.1 | - | 5, 6 | 503 | |||
AL558423.3 | - | 4, 5 | 462 | |||
AY027523.1 | - | 4, 5 | 464 | |||
AY134747.1 | - | 5, 6 | 516 | |||
BC016948.1 | - | 5, 6 | 492 | |||
BC048210.1 | - | 6, 7 | 572 | |||
BC080598.1 | - | 5, 6 | 503 | |||
BC093405.1 | - | 5, 6 | 539 | |||
BC150178.1 | - | 5, 6 | 505 | 40148074 | ||
BG778940.1 | - | 6, 7 | 609 | |||
BM906582.1 | - | 5, 6 | 528 | |||
L09749.1 | - | 4, 5 | 421 |