Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VRK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025778.1 | NP_001020949.1 | 13 | 1553 | ||
NM_016440.3 | NP_057524.3 | 14 | 1703 | |||
XM_005258971.3 | XP_005259028.1 | 14 | 2030 | |||
XM_005258972.3 | XP_005259029.1 | 13 | 1880 | |||
XM_006723237.3 | XP_006723300.1 | 14 | 1844 | |||
XM_006723238.3 | XP_006723301.1 | 13 | 1494 | |||
GenBank mRNA | AB031052.1 | - | 14 | 1475 | ||
AF514788.1 | - | 14 | 1535 | |||
AK292918.1 | - | 14 | 1446 | |||
AK294802.1 | - | 11 | 1143 | |||
AK303010.1 | - | 13 | 1328 | |||
BC095449.1 | - | 14 | 1455 | 30721912 |