Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VRK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025778.1 | NP_001020949.1 | 9 | 1101 | ||
NM_001308420.1 | NP_001295349.1 | 10 | 1089 | |||
NM_016440.3 | NP_057524.3 | 10 | 1251 | |||
XM_005258971.3 | XP_005259028.1 | 10 | 1578 | |||
XM_005258972.3 | XP_005259029.1 | 9 | 1428 | |||
XM_006723237.3 | XP_006723300.1 | 10 | 1392 | |||
XM_006723238.3 | XP_006723301.1 | 9 | 1042 | |||
XM_011527023.2 | XP_011525325.1 | 10 | 1578 | |||
GenBank mRNA | AB031052.1 | - | 10 | 1023 | ||
AF514788.1 | - | 10 | 1083 | |||
AK225154.1 | - | 10 | 996 | |||
AK292918.1 | - | 10 | 994 | |||
AK294802.1 | - | 7 | 691 | |||
AK303010.1 | - | 9 | 876 | |||
AK303529.1 | - | 9 | 927 | |||
AK311220.1 | - | 11 | 1054 | |||
BC023556.2 | - | 10 | 1030 | 4298578 | ||
BC095449.1 | - | 10 | 1003 | 30721912 |