Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRIM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016441.2 | NP_057525.1 | 13 | 2399 | ||
XM_005264357.4 | XP_005264414.1 | 12 | 2221 | |||
XM_011532898.2 | XP_011531200.1 | 14 | 2520 | |||
XM_011532899.2 | XP_011531201.1 | 14 | 2521 | |||
XM_017004258.1 | XP_016859747.1 | 13 | 2277 | |||
XM_017004259.1 | XP_016859748.1 | 13 | 2538 | |||
XM_017004260.1 | XP_016859749.1 | 12 | 2152 | |||
XM_017004261.1 | XP_016859750.1 | 12 | 1812 | |||
XM_017004262.1 | XP_016859751.1 | 11 | 1695 | |||
GenBank mRNA | AB209139.1 | - | 13 | 2399 | ||
AF167706.1 | - | 13 | 2372 | |||
AK300817.1 | - | 2 | 297 | |||
AY358371.1 | - | 13 | 2700 | |||
BC111989.1 | - | 13 | 2370 | 8327457 | ||
BC113371.1 | - | 13 | 2370 | 8322423 | ||
BX648755.1 | - | 7 | 1048 |