Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRIM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016441.2 | NP_057525.1 | 16 | 2979 | ||
XM_005264357.4 | XP_005264414.1 | 15 | 2801 | |||
XM_011532898.2 | XP_011531200.1 | 17 | 3100 | |||
XM_011532899.2 | XP_011531201.1 | 16 | 2906 | |||
XM_011532901.2 | XP_011531203.1 | 16 | 2879 | |||
XM_017004258.1 | XP_016859747.1 | 16 | 2857 | |||
XM_017004259.1 | XP_016859748.1 | 15 | 2923 | |||
XM_017004260.1 | XP_016859749.1 | 15 | 2732 | |||
XM_017004261.1 | XP_016859750.1 | 15 | 2392 | |||
XM_017004262.1 | XP_016859751.1 | 14 | 2275 | |||
GenBank mRNA | AB209139.1 | - | 16 | 2979 | ||
AF167706.1 | - | 16 | 2952 | |||
AK300817.1 | - | 5 | 781 | |||
AY358371.1 | - | 16 | 3280 | |||
BC111989.1 | - | 16 | 2950 | 8327457 | ||
BC113371.1 | - | 16 | 2950 | 8322423 | ||
BX648755.1 | - | 10 | 1628 |