Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC174 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016474.4 | NP_057558.3 | 11 | 1448 | ||
NR_135523.1 | 10 | 1353 | ||||
XM_017006554.1 | XP_016862043.1 | 9 | 1283 | |||
GenBank mRNA | AF151046.1 | - | 9 | 1186 | ||
AK091158.1 | - | 7 | 793 | |||
BC013999.2 | - | 11 | 1424 | 3936914 | ||
BC033897.1 | - | 11 | 1435 | 4812977 |