Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDZD11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016484.4 | NP_057568.1 | 3, 4 | 274 | ||
XM_011530973.2 | XP_011529275.1 | 4, 5 | 438 | |||
XM_017029568.1 | XP_016885057.1 | 3, 4 | 605 | |||
GenBank mRNA | AF151061.1 | - | 3, 4 | 244 | ||
AK024746.1 | - | 3, 4 | 242 | |||
AK311539.1 | - | 2, 3 | 136 | |||
AY358829.1 | - | 3, 4 | 480 | |||
BC012996.2 | - | 3, 4 | 213 | 3530647 | ||
BC089433.1 | - | 2, 3 | 171 | |||
BG708698.1 | - | 3, 4 | 278 | |||
BX537725.1 | - | 3, 4 | 348 | |||
CR457149.1 | - | 2, 3 | 172 |