Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C6orf203 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142468.2 | NP_001135940.1 | 2 | 789 | ||
NM_001142470.2 | NP_001135942.1 | 3 | 872 | |||
NM_016487.4 | NP_057571.1 | 2 | 263 | |||
XM_011535876.2 | XP_011534178.1 | 3 | 367 | |||
XM_011535877.2 | XP_011534179.1 | 3 | 315 | |||
XM_017010919.1 | XP_016866408.1 | 2 | 262 | |||
XM_017010920.1 | XP_016866409.1 | 2 | 755 | |||
GenBank mRNA | AF151064.1 | - | 2 | 219 | ||
AK091564.1 | - | 2 | 748 | |||
BC010899.2 | - | 1 | 165 | 4078296 | ||
BX333652.2 | - | 3 | 475 | |||
DB499205.1 | - | 2 | 263 |