Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM178B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122646.2 | NP_001116118.2 | 17 | 2667 | ||
NM_001172667.1 | NP_001166138.1 | 5 | 868 | |||
NM_016490.4 | NP_057574.2 | 5 | 986 | |||
XM_011511294.2 | XP_011509596.1 | 16 | 2595 | |||
XM_017004263.1 | XP_016859752.1 | 18 | 2832 | |||
XM_017004264.1 | XP_016859753.1 | 18 | 2793 | |||
XM_017004266.1 | XP_016859755.1 | 8 | 1240 | |||
XM_017004267.1 | XP_016859756.1 | 5 | 845 | |||
XM_017004268.1 | XP_016859757.1 | 5 | 871 | |||
GenBank mRNA | AF151068.1 | - | 5 | 826 | ||
AJ710728.1 | - | 2 | 388 | |||
AK122705.1 | - | 11 | 1546 | |||
BC038583.1 | - | 5 | 947 | |||
BC039488.1 | - | 15 | 1934 | |||
BC051379.1 | - | 5 | 986 |