Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016504.2 | NP_057588.1 | 4 | 365 | ||
GenBank mRNA | AB049647.1 | - | 4 | 340 | ||
AF151084.1 | - | 4 | 349 | |||
AK023576.1 | - | 3 | 2079 | |||
AK315758.1 | - | 4 | 366 | |||
BC001066.2 | - | 4 | 336 | 2822502 | ||
BC021986.1 | - | 4 | 350 | 4095663 | ||
BG829493.1 | - | 4 | 384 | |||
BM840463.1 | - | 4 | 366 |