Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIPOX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016518.2 | NP_057602.2 | 3 | 631 | ||
XM_017024722.1 | XP_016880211.1 | 2 | 230 | |||
GenBank mRNA | AF134593.1 | - | 3 | 348 | ||
AF136970.1 | - | 3 | 317 | |||
AK022773.1 | - | 3 | 396 | |||
AK027498.1 | - | 3 | 355 | |||
BC008960.2 | - | 3 | 631 | 2961289 | ||
BC027622.2 | - | 3 | 367 | 4797450 | ||
CR457155.1 | - | 3 | 305 |