Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016544.2 | NP_057628.1 | 5, 6 | 715 | ||
XM_017004269.1 | XP_016859758.1 | 5, 6 | 746 | |||
XM_017004270.1 | XP_016859759.1 | 5, 6 | 606 | |||
GenBank mRNA | AF178983.1 | - | 5, 6 | 549 | ||
AK294726.1 | - | 5, 6 | 499 | |||
BC034049.1 | - | 5, 6 | 576 | 4815297 |