Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198559.1 | NP_001185488.1 | 6 | 736 | ||
NM_016544.2 | NP_057628.1 | 7 | 897 | |||
XM_017004269.1 | XP_016859758.1 | 7 | 928 | |||
XM_017004270.1 | XP_016859759.1 | 7 | 788 | |||
GenBank mRNA | AF178983.1 | - | 7 | 731 | ||
AK294726.1 | - | 7 | 681 | |||
AL137731.1 | - | 9 | 1006 | |||
AY094594.1 | - | 6 | 546 | |||
BC034049.1 | - | 7 | 758 | 4815297 |