Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERGIC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016570.2 | NP_057654.2 | 3 | 317 | ||
GenBank mRNA | AF183410.1 | - | 3 | 268 | ||
AF216751.1 | - | 3 | 272 | |||
AF302767.1 | - | 3 | 270 | |||
AK074520.1 | - | 3 | 292 | |||
AK222799.1 | - | 3 | 257 | |||
AL834128.2 | - | 3 | 258 | |||
BC000887.1 | - | 3 | 270 | 3462408 | ||
BC107794.1 | - | 3 | 274 | 30319807 | ||
BF681475.1 | - | 343 | ||||
BI547753.1 | - | 3 | 321 |