Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GMIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288998.1 | NP_001275927.1 | 18 | 2148 | ||
NM_001288999.1 | NP_001275928.1 | 18 | 2157 | |||
NM_016573.3 | NP_057657.2 | 19 | 2235 | |||
XM_005259927.2 | XP_005259984.1 | 19 | 2211 | |||
XM_011528048.2 | XP_011526350.1 | 18 | 2102 | |||
XM_017026861.1 | XP_016882350.1 | 19 | 2220 | |||
XM_017026862.1 | XP_016882351.1 | 19 | 2220 | |||
GenBank mRNA | AF132541.1 | - | 19 | 2176 | ||
AK297220.1 | - | 18 | 2087 | |||
BC032057.1 | - | 19 | 2172 | |||
BC126436.1 | - | 19 | 2129 | 8992152 | ||
BC144142.1 | - | 18 | 2051 | 9052662 |