Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC15A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016582.2 | NP_057666.1 | 8 | 1852 | ||
NR_027391.1 | 7 | 1683 | ||||
XM_011545095.2 | XP_011543397.1 | 9 | 2198 | |||
GenBank mRNA | AB020598.1 | - | 8 | 1852 | ||
AF135600.1 | - | 8 | 1745 | |||
AK127216.1 | - | 3 | 4307 | |||
BC025710.1 | - | 7 | 1543 | 5224878 | ||
BC037974.1 | - | 8 | 1712 | 5213632 |