Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THEG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016585.4 | NP_057669.1 | 5 | 656 | ||
NM_199202.2 | NP_954672.1 | 4 | 584 | |||
XM_011528049.2 | XP_011526351.1 | 5 | 1335 | |||
XM_011528050.2 | XP_011526352.1 | 4 | 1262 | |||
XM_011528052.2 | XP_011526354.1 | 5 | 1334 | |||
GenBank mRNA | AB033129.1 | - | 5 | 656 | ||
AF268610.1 | - | 5 | 622 | |||
BC028574.2 | - | 5 | 635 | 4821678 |