Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM53C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135647.1 | NP_001129119.1 | 4, 5 | 1389 | ||
NM_016605.2 | NP_057689.1 | 4, 5 | 1364 | |||
GenBank mRNA | AF251040.1 | - | 4, 5 | 1100 | ||
AK027412.1 | - | 2, 3 | 255 | |||
AK310378.1 | - | 4, 5 | 542 | |||
AK315567.1 | - | 4, 5 | 1113 | |||
BC000259.1 | - | 4, 5 | 1106 | 3357346 | ||
BC052993.1 | - | 4, 5 | 1329 | 5164799 |