Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REEP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271803.1 | NP_001258732.1 | 2 | 287 | ||
NM_016606.3 | NP_057690.2 | 2 | 287 | |||
NR_073448.1 | 2 | 362 | ||||
NR_073449.1 | 2 | 362 | ||||
GenBank mRNA | AF251041.1 | - | 1 | 60 | ||
AK056193.1 | - | 2 | 243 | |||
AK223611.1 | - | 2 | 228 | |||
AK290547.1 | - | 2 | 177 | |||
AK291101.1 | - | 2 | 240 | |||
AK293480.1 | - | 2 | 261 | |||
AY562240.1 | - | 2 | 79 | |||
BC006218.2 | - | 2 | 201 | 3940709 | ||
DC424086.1 | - | 2 | 313 |