Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289050.1 | NP_001275979.1 | 1 | 455 | ||
NM_016609.4 | NP_057693.3 | 2 | 722 | |||
NM_020372.3 | NP_065105.2 | 1 | 455 | |||
NR_110290.1 | 2 | 722 | ||||
XM_005267747.4 | XP_005267804.1 | 2 | 2798 | |||
XM_005267748.4 | XP_005267805.1 | 2 | 2801 | |||
XM_017021361.1 | XP_016876850.1 | 2 | 2795 | |||
XM_017021362.1 | XP_016876851.1 | 2 | 2793 | |||
GenBank mRNA | AK308661.1 | - | 1 | 580 | ||
AK311241.1 | - | 1 | 47 | |||
BC020565.1 | - | 1 | 274 | 4156259 | ||
BX161416.1 | - | 2 | 681 | |||
DQ658848.1 | - | 1 | 225 |