Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDH9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016279.3 | NP_057363.3 | 3 | 470 | ||
GenBank mRNA | AB035302.1 | - | 3 | 455 | ||
AK307779.1 | - | 3 | 470 | |||
AK308697.1 | - | 3 | 474 | |||
BC107594.1 | - | 3 | 432 | 40011942 | ||
BC113745.1 | - | 3 | 432 | 40011941 | ||
DA165253.1 | - | 3 | 470 |