Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDH9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016279.3 | NP_057363.3 | 12 | 2135 | ||
GenBank mRNA | AB035302.1 | - | 12 | 2120 | ||
AK294183.1 | - | 5 | 982 | |||
AK307779.1 | - | 12 | 2136 | |||
BC107594.1 | - | 12 | 2097 | 40011942 | ||
BC113745.1 | - | 12 | 2097 | 40011941 |