Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSRC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271834.1 | NP_001258763.1 | 2 | 297 | ||
NM_001271838.1 | NP_001258767.1 | 2 | 323 | |||
NM_016625.3 | NP_057709.2 | 2 | 311 | |||
GenBank mRNA | AF208853.1 | - | 2 | 269 | ||
AK290334.1 | - | 2 | 291 | |||
AK300388.1 | - | 2 | 207 | |||
BC006982.1 | - | 2 | 260 | 3997840 | ||
BC010357.1 | - | 2 | 246 | 4249317 | ||
DA273191.1 | - | 2 | 313 |