Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AHSP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318221.1 | NP_001305150.1 | 3 | 495 | ||
NM_001318222.1 | NP_001305151.1 | 3 | 426 | |||
NM_016633.3 | NP_057717.1 | 3 | 499 | |||
GenBank mRNA | AF112995.1 | - | 3 | 530 | ||
AF147435.1 | - | 3 | 458 | |||
AF208865.1 | - | 3 | 455 | |||
AF364517.1 | - | 3 | 455 | |||
BC035842.1 | - | 3 | 440 | 5748040 |