Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZMYND10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308379.1 | NP_001295308.1 | 6 | 735 | ||
NM_015896.3 | NP_056980.2 | 6 | 735 | |||
XM_005265216.2 | XP_005265273.1 | 5 | 908 | |||
GenBank mRNA | AB209621.1 | - | 1 | 3378 | ||
AK058135.1 | - | 6 | 665 | |||
AK093455.1 | - | 1 | 20 | |||
AK096525.1 | - | 6 | 1843 | |||
AK223343.1 | - | 6 | 650 | |||
AK308086.1 | - | 6 | 634 | |||
AK315123.1 | - | 6 | 721 | |||
AL833828.1 | - | 6 | 664 | |||
BC033732.1 | - | 6 | 631 | 5168611 | ||
U70824.1 | - | 6 | 689 | |||
U70880.1 | - | 6 | 689 |