Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDH13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001220488.1 | NP_001207417.1 | 5, 6 | 892 | ||
NM_001220490.1 | NP_001207419.1 | 3, 4 | 577 | |||
NM_001257.4 | NP_001248.1 | 4, 5 | 786 | |||
XM_011522804.2 | XP_011521106.1 | 3, 4 | 393 | |||
XM_017022848.1 | XP_016878337.1 | 5, 6 | 809 | |||
GenBank mRNA | AK295886.1 | - | 3, 4 | 407 | ||
AK307862.1 | - | 5, 6 | 583 | |||
BC028624.2 | - | 4, 5 | 578 | 4825734 | ||
BC030653.2 | - | 4, 5 | 578 | 4811561 | ||
BI756586.1 | - | 4, 5 | 589 | |||
CV024366.1 | - | 4, 5 | 487 | |||
L34058.1 | - | 4, 5 | 931 | |||
U59288.1 | - | 4, 5 | 703 |