Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015976.4 | NP_057060.2 | 2 | 299 | ||
NM_152238.3 | NP_689424.2 | 2 | 299 | |||
NR_033716.1 | 3 | 419 | ||||
XM_006710677.2 | XP_006710740.1 | 2 | 316 | |||
XM_006710678.2 | XP_006710741.1 | 3 | 415 | |||
XM_011541564.2 | XP_011539866.1 | 3 | 355 | |||
XM_017001425.1 | XP_016856914.1 | 2 | 277 | |||
XM_017001426.1 | XP_016856915.1 | 3 | 414 | |||
XM_017001427.1 | XP_016856916.1 | 3 | 689 | |||
XM_017001428.1 | XP_016856917.1 | 2 | 164 | |||
GenBank mRNA | AF121857.1 | - | 2 | 142 | ||
AK025031.1 | - | 2 | 256 | |||
AK223229.1 | - | 2 | 158 | |||
AK293018.1 | - | 2 | 227 | |||
AK298211.1 | - | 2 | 145 | |||
AL049989.1 | - | 2 | 156 | |||
AL548028.3 | - | 2 | 300 | |||
BC010349.1 | - | 2 | 113 | 4093672 | ||
BC018105.1 | - | 3 | 237 | 3849824 | ||
BI856196.1 | - | 2 | 320 | |||
BT006747.1 | - | 1 | 55 |